2-(4-Chlorophenylthio)nicotinic acid
Supplier: Thermo Scientific
BTB08734.10
BTB08734.50
MAYBBTB08734.10EA
0
EUR
MAYBBTB08734.10
MAYBBTB08734.50
2-(4-Chlorophenylthio)nicotinic acid
2-(4-Chlorophenylthio)nicotinic acid
Formula:
C₁₂H₈ClNO₂S |
MDL Number:
MFCD00052117 CAS Number: 955-54-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...