EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate), Sigma-Aldrich®
Supplier: Merck
Synonyms: 2-Ethoxy-1-ethoxycarbonyl-1,2-dihydroquinoline, N-Ethoxycarbonyl-2-ethoxy-1,2-dihydroquinoline, Ethyl 2-ethoxyquinoline-1(2H)-carboxylate
M-Clarity™ quality level = MQ200
149837-100G 149837-25G 149837-5G
SIAL149837-100GEA 126 EUR
SIAL149837-100G SIAL149837-25G SIAL149837-5G
EEDQ (ethyl-N-(2-ethoxy-1,2-dihydroquinoline)carboxylate), Sigma-Aldrich®
EEDQ (Ethyl N-(2-ethoxy-1,2-dihydroquinoline)carboxylate)
Formula: C₁₄H₁₇NO₃
MW: 247.29 g/mol
MDL Number: MFCD00006703
CAS Number: 16357-59-8
EINECS: 240-418-2