3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonitrile

Supplier: Thermo Scientific

RF00723.1 RF00723.10 RF00723.50
MAYBRF00723.1EA 0 EUR
MAYBRF00723.1 MAYBRF00723.10 MAYBRF00723.50
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonitrile
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carbonitrile
Formula: C₁₁H₆Cl₂N₂O
MDL Number: MFCD00052552
CAS Number: 21486-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR