Dihydrojasmone, Sigma-Aldrich®

Supplier: Merck

Synonyms: 3-Methyl-2-pentylcyclopent-2-en-1-one

M-Clarity™ quality level = MQ300

W376302-100G-K W376302-1KG-K W376302-5KG-K
SIALW376302-100G-KEA 80.5 EUR
SIALW376302-100G-K SIALW376302-1KG-K SIALW376302-5KG-K
Dihydrojasmone, Sigma-Aldrich®
Dihydrojasmone
Formula: C₁₁H₁₈O
MDL Number: MFCD00036480
CAS Number: 1128-08-1
EINECS: 214-434-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR