Methyl dimethoxyacetate 98%
Supplier: Thermo Fisher Scientific
Synonyms: Methyl 2,2-dimethoxyacetate
414180250 414180050
ACRO414180250EA 123 EUR
ACRO414180250 ACRO414180050
Methyl dimethoxyacetate 98%
Methyl dimethoxyacetate
Formula: C₅H₁₀O₄
MW: 134.13 g/mol
Boiling Pt: 160…163 °C (760 mmHg)
Density: 1.09 g/cm³
Flash Pt: 63 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008484
CAS Number: 89-91-8
EINECS: 201-950-0
Order Now
Specification Test Results
Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4040 to 1.4070 (20°C, 589 nm)
GC ≥97.5 %