Palladium(II) nitrate 10% in nitric acid 10%, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
380040-10ML
380040-50ML
SIAL380040-10MLEA
140
EUR
SIAL380040-10ML
SIAL380040-50ML
Palladium(II) nitrate 10% in nitric acid 10%, Sigma-Aldrich®
Palladium (II) nitrate
Formula:
N₂O₆Pd MW: 230.43 g/mol |
MDL Number:
MFCD00011169 CAS Number: 10102-05-3 EINECS: 233-265-8 UN: 3264 ADR: ,II |