Empty solid phase extraction cartridges with two frits, Bond Elut™
Supplier: VARIAN
12131014 12131015 12131016 12131017 12131018 12131013 12131014S
551-0362EA 0 EUR
551-0362 551-0363 551-0364 551-0365 551-0366 VARI12131013 VARI12131014S
Empty solid phase extraction cartridges with two frits, Bond Elut™
Chromatography Columns
Clean PP reservoirs contain two PP frits pre-inserted, an ideal configuration for simple filtration.
  • Pre-installed frits for ease-of-use
  • Broad range of filtration operations for maximum flexibility
  • Customisable packing for specific applications