Anti-GABA Transporter 3 / GAT 3 Rabbit Monoclonal Antibody [clone: EPR25153-38]
Supplier: Abcam
AB300559-100 AB300559-10 AB300559-20UL AB300559-1ML
ABCAAB300559-100EA 707 EUR
ABCAAB300559-100 ABCAAB300559-10 ABCAAB300559-20UL ABCAAB300559-1ML
Anti-GABA Transporter 3 / GAT 3 Rabbit Monoclonal Antibody [clone: EPR25153-38]
Antibodies
Rabbit monoclonal [EPR25153-38] to GABA Transporter 3 / GAT 3.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: GABA Transporter 3 / GAT 3
Clonality: Monoclonal
Clone: EPR25153-38
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat