Anti-LIME Rabbit Monoclonal Antibody [clone: EPR14713]

Supplier: Abcam
AB188871-100 AB188871-1ML
ABCAAB188871-100EA 638 EUR
ABCAAB188871-100 ABCAAB188871-1ML
Anti-LIME Rabbit Monoclonal Antibody [clone: EPR14713]
Antibodies
Rabbit monoclonal [EPR14713] to LIME.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LIME
Clonality: Monoclonal
Clone: EPR14713
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR