Anti-Glutamine Synthetase Rabbit Monoclonal Antibody [clone: EPR13022(B)] (Alexa Fluor® 594)
Supplier: Abcam
AB305118-100 AB305118-10
ABCAAB305118-100EA 775 EUR
ABCAAB305118-100 ABCAAB305118-10
Anti-Glutamine Synthetase Rabbit Monoclonal Antibody [clone: EPR13022(B)] (Alexa Fluor® 594)
Antibodies
Alexa Fluor® 594 Rabbit monoclonal [EPR13022(B)] to Glutamine Synthetase.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Glutamine Synthetase
Clonality: Monoclonal
Clone: EPR13022(B)
Conjugation: Alexa Fluor® 594
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human