Anti-ADAMTS1 Rabbit Monoclonal Antibody [clone: EPR23324-51]
Supplier: Abcam
AB281042-1 AB281042-100
ABCAAB281042-1EA 8680 EUR
ABCAAB281042-1 ABCAAB281042-100
Anti-ADAMTS1 Rabbit Monoclonal Antibody [clone: EPR23324-51]
Antibodies
Rabbit monoclonal [EPR23324-51] to ADAMTS1 - BSA and Azide free (Detector).
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ADAMTS1
Clonality: Monoclonal
Clone: EPR23324-51
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat