Anti-CCR6 Rabbit Monoclonal Antibody [clone: EPR24085-145]

Supplier: Abcam
AB303672-100 AB303672-20UL AB303672-1ML
ABCAAB303672-100EA 707 EUR
ABCAAB303672-100 ABCAAB303672-20UL ABCAAB303672-1ML
Anti-CCR6 Rabbit Monoclonal Antibody [clone: EPR24085-145]
Antibodies
Rabbit monoclonal [EPR24085-145] to CCR6.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: EPR24085-145
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR