Anti-Dopamine Transporter Rabbit Monoclonal Antibody [clone: EPR19695] (Alexa Fluor® 568)
Supplier: Abcam
AB303466-100 AB303466-10
ABCAAB303466-100EA 775 EUR
ABCAAB303466-100 ABCAAB303466-10
Anti-Dopamine Transporter Rabbit Monoclonal Antibody [clone: EPR19695] (Alexa Fluor® 568)
Antibodies
Alexa Fluor® 568 Rabbit monoclonal [EPR19695] to Dopamine Transporter.
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Dopamine Transporter
Clonality: Monoclonal
Clone: EPR19695
Conjugation: Alexa Fluor® 568
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat