Anti-Lactate Dehydrogenase Rabbit Monoclonal Antibody [clone: EP1566Y]
Supplier: Abcam
AB219591-100 AB219591-1
ABCAAB219591-100EA 1020 EUR
ABCAAB219591-100 ABCAAB219591-1
Anti-Lactate Dehydrogenase Rabbit Monoclonal Antibody [clone: EP1566Y]
Antibodies
Rabbit monoclonal [EP1566Y] to Lactate Dehydrogenase - BSA and Azide free.
Type: Primary
Antigen: Lactate Dehydrogenase
Clonality: Monoclonal
Clone: EP1566Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat; Human