Anti-Tissue Factor Rabbit Monoclonal Antibody [clone: EPR22548-240]

Supplier: Abcam
AB252918-100 AB252918-20UL AB252918-1ML
ABCAAB252918-100EA 738 EUR
ABCAAB252918-100 ABCAAB252918-20UL ABCAAB252918-1ML
Anti-Tissue Factor Rabbit Monoclonal Antibody [clone: EPR22548-240]
Antibodies
Rabbit monoclonal [EPR22548-240] to Tissue Factor.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Tissue Factor
Clonality: Monoclonal
Clone: EPR22548-240
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR