Anti-PTP zeta/ Phosphacan Rabbit Monoclonal Antibody [clone: EPR26270-86]

Supplier: Abcam
AB290640-100 AB290640-20UL AB290640-1ML
ABCAAB290640-100EA 707 EUR
ABCAAB290640-100 ABCAAB290640-20UL ABCAAB290640-1ML
Anti-PTP zeta/ Phosphacan Rabbit Monoclonal Antibody [clone: EPR26270-86]
Antibodies
Rabbit monoclonal [EPR26270-86] to PTP zeta/ Phosphacan.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: PTP zeta/ Phosphacan
Clonality: Monoclonal
Clone: EPR26270-86
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR