Anti-Dectin-1 Rabbit Monoclonal Antibody [clone: EPR25360-64]
Supplier: Abcam
AB287961-100 AB287961-20UL AB287961-1ML
ABCAAB287961-100EA 675 EUR
ABCAAB287961-100 ABCAAB287961-20UL ABCAAB287961-1ML
Anti-Dectin-1 Rabbit Monoclonal Antibody [clone: EPR25360-64]
Antibodies
Rabbit monoclonal [EPR25360-64] to Dectin-1.
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Dectin-1
Clonality: Monoclonal
Clone: EPR25360-64
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse