Anti-HIV1 p24+HIV1 p17+HIV1 p55 Rabbit Recombinant Multiclonal Antibody [clone: RM2002]

Supplier: Abcam
AB309160-1MG AB309160-100UG
ABCAAB309160-1MGEA 9160 EUR
ABCAAB309160-1MG ABCAAB309160-100UG
Anti-HIV1 p24+HIV1 p17+HIV1 p55 Rabbit Recombinant Multiclonal Antibody [clone: RM2002]
Antibodies
Rabbit recombinant multiclonal [RM2002] to HIV1 p24 + HIV1 p17 + HIV1 p55 - BSA and Azide free.

Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: HIV1 p24+HIV1 p17+HIV1 p55
Clonality: Recombinant Multiclonal
Clone: RM2002
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: HIV
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR