Anti-DES Rabbit Monoclonal Antibody [clone: SP138]
Supplier: Abcam
AB227651-100 AB227651-1 AB227651-500
ABCAAB227651-100EA 732 EUR
ABCAAB227651-100 ABCAAB227651-1 ABCAAB227651-500
Anti-DES Rabbit Monoclonal Antibody [clone: SP138]
Antibodies
Rabbit monoclonal [SP138] to Desmin - C-terminal.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: DES
Clonality: Monoclonal
Clone: SP138
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat