Anti-PKA alpha/beta/gamma (catalytic subunit) Rabbit Monoclonal Antibody [clone: EP2606Y]

Supplier: Abcam
AB227848-100 AB227848-1
ABCAAB227848-100EA 1020 EUR
ABCAAB227848-100 ABCAAB227848-1
Anti-PKA alpha/beta/gamma (catalytic subunit) Rabbit Monoclonal Antibody [clone: EP2606Y]
Antibodies
Rabbit monoclonal [EP2606Y] to PKA alpha/beta/gamma (catalytic subunit) (phospho T197) - BSA and Azide free.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PKA alpha/beta/gamma (catalytic subunit)
Clonality: Monoclonal
Clone: EP2606Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Rat;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR