Anti-BRAF Rat Monoclonal Antibody [clone: pBR1]

Supplier: Abcam
AB183337-100 AB183337-500 AB183337-1
ABCAAB183337-100EA 694 EUR
ABCAAB183337-100 ABCAAB183337-500 ABCAAB183337-1
Anti-BRAF Rat Monoclonal Antibody [clone: pBR1]
Antibodies
Rat monoclonal [pBR1] to BRAF.

Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: BRAF
Clonality: Monoclonal
Clone: pBR1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR