Anti-Noradrenaline transporter Rabbit Monoclonal Antibody [clone: EPR23527-534]
Supplier: Abcam
AB282022-1 AB282022-100
ABCAAB282022-1EA 9160 EUR
ABCAAB282022-1 ABCAAB282022-100
Anti-Noradrenaline transporter Rabbit Monoclonal Antibody [clone: EPR23527-534]
Rabbit monoclonal [EPR23527-534] to Noradrenaline transporter - BSA and Azide free.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Noradrenaline transporter
Clonality: Monoclonal
Clone: EPR23527-534
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human;Rat