Anti-MCT1/Monocarboxylic acid transporter 1 Rabbit Monoclonal Antibody [clone: EPR26702-83] (Alexa Fluor® 488)
Supplier: Abcam
AB321856-10UL AB321856-100UL
ABCAAB321856-10ULEA 238 EUR
ABCAAB321856-10UL ABCAAB321856-100UL
Anti-MCT1/Monocarboxylic acid transporter 1 Rabbit Monoclonal Antibody [clone: EPR26702-83] (Alexa Fluor® 488)
Antibodies
Alexa Fluor® 488 Rabbit monoclonal [EPR26702-83] to MCT1/Monocarboxylic acid transporter 1.
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: MCT1/Monocarboxylic acid transporter 1
Clonality: Monoclonal
Clone: EPR26702-83
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human