Anti-MCT1/Monocarboxylic acid transporter 1 Rabbit Monoclonal Antibody [clone: EPR26702-83]

Supplier: Abcam
AB315382-20UL AB315382-100UL AB315382-1ML
ABCAAB315382-20ULEA 244 EUR
ABCAAB315382-20UL ABCAAB315382-100UL ABCAAB315382-1ML
Anti-MCT1/Monocarboxylic acid transporter 1 Rabbit Monoclonal Antibody [clone: EPR26702-83]
Antibodies
Rabbit monoclonal [EPR26702-83] to MCT1/Monocarboxylic acid transporter 1.

Type: Primary
Antigen: MCT1/Monocarboxylic acid transporter 1
Clonality: Monoclonal
Clone: EPR26702-83
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR