Anti-eNOS Rabbit Recombinant Multiclonal Antibody [clone: RM1181]
Supplier: Abcam
AB317583-100UG AB317583-1MG
ABCAAB317583-100UGEA 1020 EUR
ABCAAB317583-100UG ABCAAB317583-1MG
Anti-eNOS Rabbit Recombinant Multiclonal Antibody [clone: RM1181]
Antibodies
Rabbit recombinant multiclonal [RM1181] to eNOS - BSA and Azide free.
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: eNOS
Clonality: Recombinant Multiclonal
Clone: RM1181
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat