Anti-Tissue Plasminogen Activator Rabbit Monoclonal Antibody [clone: EPR7232(2)] (Biotinylated)
Supplier: Abcam
AB314267-10UL AB314267-100UL
ABCAAB314267-10ULEA 238 EUR
ABCAAB314267-10UL ABCAAB314267-100UL
Anti-Tissue Plasminogen Activator Rabbit Monoclonal Antibody [clone: EPR7232(2)] (Biotinylated)
Antibodies
Biotin Rabbit monoclonal [EPR7232(2)] to Tissue Plasminogen Activator.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Tissue Plasminogen Activator
Clonality: Monoclonal
Clone: EPR7232(2)
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human