Anti-RanBP2 Rabbit Monoclonal Antibody [clone: EPR28204-184]
Supplier: Abcam
AB315458-20UL AB315458-100UL AB315458-1ML
ABCAAB315458-20ULEA 244 EUR
ABCAAB315458-20UL ABCAAB315458-100UL ABCAAB315458-1ML
Anti-RanBP2 Rabbit Monoclonal Antibody [clone: EPR28204-184]
Antibodies
Rabbit monoclonal [EPR28204-184] to RanBP2.
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: RanBP2
Clonality: Monoclonal
Clone: EPR28204-184
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human