Anti-Lipocalin-2/NGAL Rabbit Recombinant Multiclonal Antibody [clone: RM1211]
Supplier: Abcam
AB318209-20UL AB318209-100UL AB318209-1ML
ABCAAB318209-20ULEA 207 EUR
ABCAAB318209-20UL ABCAAB318209-100UL ABCAAB318209-1ML
Anti-Lipocalin-2/NGAL Rabbit Recombinant Multiclonal Antibody [clone: RM1211]
Antibodies
Rabbit recombinant multiclonal [RM1211] to Lipocalin-2 / NGAL.
Upozornení: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Lipocalin-2 / NGAL
Clonality: Recombinant Multiclonal
Clone: RM1211
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat