Anti-MCT1/Monocarboxylic acid transporter 1 Rabbit Monoclonal Antibody [clone: EPR26702-83] (Alexa Fluor® 647)
Supplier: Abcam
AB320781-10UL AB320781-100UL
ABCAAB320781-10ULEA 238 EUR
ABCAAB320781-10UL ABCAAB320781-100UL
Anti-MCT1/Monocarboxylic acid transporter 1 Rabbit Monoclonal Antibody [clone: EPR26702-83] (Alexa Fluor® 647)
Antibodies
Alexa Fluor® 647 Rabbit monoclonal [EPR26702-83] to MCT1/Monocarboxylic acid transporter 1.
Upozornení: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MCT1/Monocarboxylic acid transporter 1
Clonality: Monoclonal
Clone: EPR26702-83
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human