phynomic x-foam HV Safety Gloves, 60054 Series
Supplier: UVEX
6005411 6005412
UVEX6005411EA 76 EUR
UVEX6005411 UVEX6005412
phynomic x-foam HV Safety Gloves, 60054 Series
Gloves
These safety gloves fit like a second skin and are particularly light and flexible, without making the hands sweat.
  • Unique safety glove with break sections
  • Extremely good skin compatibility and free from hazardous substances
  • Aqua-polymer foam coating on the palm and fingertips
A seamless break section is integrated into the fingers of the uvex phynomic x-foam HV, significantly reducing the tear resistance and thus hugely reducing the risk of serious hand injuries when using manually operated screwdrivers.