rubiflex S NB35S Chemical Protection Gloves
Supplier: UVEX
6019001 6019002 6019003
UVEX6019001EA 508 EUR
UVEX6019001 UVEX6019002 UVEX6019003
rubiflex S NB35S Chemical Protection Gloves
Gloves
These chemical protection gloves feature a thicker cotton interlock lining. This makes the uvex rubiflex S suitable for many applications in which protection against chemicals and increased mechanical protection (for example against heat) is required.
  • NBR chemical protection glove with reinforced cotton interlock supporting material
  • Fully coated with special NBR coating (nitrile rubber), approx. 0.50 mm thick
  • Excellent chemical resistance and food-safe
  • Flexible
The rubiflex S chemical protection gloves are characterised by excellent wearer comfort, good mechanical abrasion resistance, and good resistance to many chemicals, acids, alkalis, mineral oils, and solvents. Their ergonomic fit provides a good tactile feel and flexibility.