unipur Carbon FT Assembly Gloves, 60587 Series
Supplier: UVEX
6058786 6058787 6058788 6058789 6058790 6058791
UVEX6058786EA 39.2 EUR
UVEX6058786 UVEX6058787 UVEX6058788 UVEX6058789 UVEX6058790 UVEX6058791
unipur Carbon FT Assembly Gloves, 60587 Series
Gloves
The unipur carbon FT glove is suited to precision mechanical work in antistatic work environments, while also offering an exceptional tactile feel.
  • Thin elastomer coating on fingertips
  • Very high breathability
  • Outstanding tactivle feel
Sensitive and anti-static safety glove for precision work with electronic parts.