phynomic wet Safety Gloves, 60060 Series
Supplier: UVEX
6006006 6006011 6006012
UVEX6006006EA 60.1 EUR
UVEX6006006 UVEX6006011 UVEX6006012
phynomic wet Safety Gloves, 60060 Series
Gloves
The phynomic wet safety glove is designed for optimal performance in damp and wet conditions.
  • Good grip in damp or wet conditions
  • Very high abrasion resistance
  • Excellent skin tolerance and free from pollutants
Featuring an aqua-polymer foam coating on the palm and fingertips, these gloves provide excellent grip and mechanical abrasion resistance. The lightweight and flexible design ensures a comfortable fit, making them feel like a second skin. With high breathability and outstanding tactile sensitivity, these gloves are ideal for precision assembly work. Additionally, they are dermatologically tested and free from harmful substances, ensuring excellent skin compatibility.