Monkey CRP ELISA Kits (C-Reactive protein)
Supplier: Abcam
AB260062-1X96 AB260062-10X96 AB260062-1X384
ABCAAB260062-1X96EA 975 EUR
ABCAAB260062-1X96 ABCAAB260062-10X96 ABCAAB260062-1X384
Monkey CRP ELISA Kits (C-Reactive protein)
Assays ELISAs
Monkey CRP ELISA Kit (C-Reactive protein) is a single-wash 90-min Simplestep used to quantify Monkey CRP (C-Reactive protein) with a sensitivity of 9,14 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput - Validated on a number of sample types including cerebrospinal fluid (CSF).