Human Tissue Kallikrein (KLK1) ELISA Kits- Intracellular
Supplier: Abcam
AB320796-10X96 AB320796-1X384 AB320796-96TST
ABCAAB320796-10X96EA 8160 EUR
ABCAAB320796-10X96 ABCAAB320796-1X384 ABCAAB320796-96TST
Human Tissue Kallikrein (KLK1) ELISA Kits- Intracellular
Assays ELISAs
Human Tissue Kallikrein (KLK1) ELISA kit- Intracellular is a single-wash 90-min Simplestep used to quantify Human Tissue Kallikrein (KLK1) with a sensitivity of 4.8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput .