Human Tissue Kallikrein (KLK1) ELISA Kits- Intracellular
Supplier: Abcam
Human Tissue Kallikrein (KLK1) ELISA kit- Intracellular is a single-wash 90-min Simplestep used to quantify Human Tissue Kallikrein (KLK1) with a sensitivity of 4.8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput .