HyFlex® 11-644 Cut Resistant Gloves
Supplier: Ansell
11-645/5 11-645/10 11-645/11 11-645/12 11-645/6 11-645/7 11-645/8 11-645/9
ANSE11-645/5EA 233 EUR
ANSE11-645/5 ANSE11-645/10 ANSE11-645/11 ANSE11-645/12 ANSE11-645/6 ANSE11-645/7 ANSE11-645/8 ANSE11-645/9
HyFlex® 11-644 Cut Resistant Gloves
Gloves
Mechanical protection gloves granting superior cut resistance and optimal comfort
  • INTERCEPT™ Cut resistance technology: Twice the cut resistance
  • Lighter fit and feel: Enhanced comfort for extended use
  • PU coating: High durability for dry and slightly oily applications
HyFlex® 11-645 protective gloves leverage INTERCEPT™ Cut Resistance Technology to deliver 2× greater cut resistance. These cut-resistant gloves provide the fit and feel of lighter protection levels. Owing to their skin-friendly PU coating, these hand protection gloves afford superb durability for dry and slightly oily applications.
In addition, these HyFlex® gloves allow wearer to use multiple touchscreen devices without having to remove their gloves. As these industrial gloves are silicone-free, they prevent contaminant transfer to metal before painting. HyFlex® 11-645 polyurethane gloves also possess Dermatest® and Oekotex certification.
Recommended for Assembly of sharp parts, handling metal sheets and panels, maintenance and inspection (picking, checking and marking).
Certifikace:
Meets ANSI/ISEA 105-2024 CUT A4 and EN ISO D safety standards.
Also possess Dermatest® and Oekotex certification, EN388: 2016, 4×43D, ANSI ABR 4, ANSI PUN 3. Category II.