NEXTFLEX™ Unique Dual Index Barcodes (Set of 1536)
Supplier: BIOO SCIENTIFIC
NOVA-534100 NOVA-534101 NOVA-534102 NOVA-534103 NOVA-534104 634100-EVAL16 634100-EVAL48 NOVA-634100 NOVA-634101 NOVA-634102 NOVA-634103 NOVA-634104 NOVA-634105 NOVA-634106 NOVA-634107 NOVA-634108 NOVA-634109 NOVA-634110 NOVA-634111 NOVA-634112 NOVA-634113 NOVA-634114 NOVA-634115 NOVA-634116
BIOONOVA-534100EA 0 EUR
BIOONOVA-534100 BIOONOVA-534101 BIOONOVA-534102 BIOONOVA-534103 BIOONOVA-534104 BIOO634100-EVAL16 BIOO634100-EVAL48 BIOONOVA-634100 BIOONOVA-634101 BIOONOVA-634102 BIOONOVA-634103 BIOONOVA-634104 BIOONOVA-634105 BIOONOVA-634106 BIOONOVA-634107 BIOONOVA-634108 BIOONOVA-634109 BIOONOVA-634110 BIOONOVA-634111 BIOONOVA-634112 BIOONOVA-634113 BIOONOVA-634114 BIOONOVA-634115 BIOONOVA-634116
NEXTFLEX™ Unique Dual Index Barcodes (Set of 1536)
Nucleic Acid Reagents Next Generation Sequencing Reagents
The NEXTFLEX® UDI Barcodes (1 to 1536) are 1536 unique dual index adapters with 10 nt indexes for multiplexing of NGS libraries compatible with Illumina® sequencers.
  • 384 UDI adapters for mitigating sample misassignment due to index hopping
  • Unique dual index adapters for multiplexing up to 384 libraries for Illumina® sequencing platforms
  • Mitigate index hopping or spread of signal that can occur during sequencing
  • Each lot of the NEXTFLEX® unique dual index barcodes is tested for index purity by sequencing
  • Compatible with both paired-end and single-read sequencing
  • Compatible with the NEXTFLEX® universal blockers
The NEXTFLEX unique dual index barcodes utilize two unique indexes on each UDI adapter, with a hamming distance of at least three throughout the entire series to permit error correction.
Each NEXTFLEX unique dual index barcode comprises the entire flow cell binding site, sequencing primer binding site and index sequences, thereby making PCR enrichment optional. Patterned flow cells, such as those found on the Illumina NovaSeq®, HiSeq® 3000/4000, and HiSeq X platforms, have been known to suffer from increased rates of sample mis-assignment during sequencing.
The use of adapters with unique dual indices improves data integrity by correctly removing mis-assigned reads. Every lot is tested for index purity and functionality by sequencing. The design of the NEXTFLEX unique dual index barcodes allow a broad range of multiplexing - users can multiplex anywhere between two samples and 1536 samples in a single sequencing run.
Certifikace: Manufactured in ISO 9001 facility.
Upozornení: For research use only. Not for use in diagnostic procedures.