jetCRISPR™ - RNP transfection reagent for efficient genome editing
jetCRISPR™ is a RNP delivery reagent, specially designed for co-delivery of guide RNA and cas9 protein for gene editing experiments. jetCRISPR™ leads to high genome editing efficiency while maintaining an excellent cell viability and morphology.
- Specifically designed for Cas9 protein and guide RNA delivery
- High genome editing efficiency with SpCas9 Nuclease
- Excellent cell viability and morphology
- Fast and reliable gene editing
- Easy to use: reverse and forward protocols