Masterflex Connection Newsletter Vol.2-4 - Robotic Dispenser System
The Masterflex robotic dispenser system includes the robotic dispenser 77700-00, 6-nozzle filling arm 77700-67, Type 1(X) guide 77700-61 for 13-mm tubes, and Type 2(Y) guide 77700-62 for 13-mm tubes, and power supply. Order Masterflex pump separately.
Improve Productivity!
NEW Robotic Dispenser System
- Fills a variety of vessels automatically
- Uses standard racks
- No complicated programming
Save time, money, and make things a lot more convenientwith our new benchtop robotic dispensing system that automatically fills a large number of vessels quickly and accurately.
Used with a Masterflex® peristaltic tubing pump, this cost-effective modular system eliminates time-consuming manual procedures and greatly enhances efficiency.
Whether for single or repetitive dispensing, it's perfect for speeding up processes for a various applications, such as media preparation, serial dilution of food samples, and small batch pharmaceutical production.
The unit measures 35" L x 10" W x 12" H (900 mm x 260 mm x 300 mm). Shipping weight is approximately 17 lb (8 kg).For additional information, please call our Application Specialists at 1-800-627-8373 or visit us online to order.