Corning® MITO+ Serum Extender, Corning
Supplier: Corning
355006
734-1317EA 196.67 EUR
734-1317
Corning® MITO+ Serum Extender, Corning
Cell Culture Supplements and Additives
MITO+ Serum Extender is a concentrated, fully defined formulation of hormones, growth factors (EGF and FGF), and other metabolites (insulin and steriod hormones).
It can be used to culture a variety of cells under serum-free or serum-reduced conditions.
Tested for ability to promote proliferation of BALB/c-3T3 cells in serum-reduced medium. Filtered (0.2 µm membrane) and tested and found negative for the presence of bacteria, fungi and mycoplasma.
(5ml; 5L equivalent).