Kits: the mystery of the crooked cell, Ready-to-Load™
Supplier: EDVOTEK
S-53
763-0010EA 82.5 EUR
763-0010
Kits: the mystery of the crooked cell, Ready-to-Load™
Educational Materials Biology Educational Materials Genetics Learning Activities
This simple experiment demonstrates detection of the mutation that causes Sickle Cell Anaemia. In this simulation, students use electrophoresis to separate dyes that represent patient samples and controls.
  • For 10 lab groups
  • Complete in 45 minutes
Also required: Electrophoresis tank and power supply.
Delivery information: Kit includes instructions, Ready-to-Load™ dye samples, agarose powder, practice gel loading solution, electrophoresis buffer, and microtipped transfer pipettes.