Experimentation kits, protein electrophoresis
Supplier: G-Biosciences
BE-406
BE-406EA 319 EUR
BE-406
Experimentation kits, protein electrophoresis
Educational Materials Biology Educational Materials Biochemistry Learning Activities
Using the Protein Electrophoresis kit, students will learn the principles of various types of electrophoresis, including denaturing and non-denaturing electrophoresis, and how this powerful technique is used to analyse proteins. The kit will introduce students to the different separation matrices currently in use and will understand their differing separation properties and their role in protein analysis. Students have an option of casting their own electrophoresis gels using polyacrylamide or using pre-cast commercially available gels. This kit is provided with all of the reagents, buffersMore Product Information