Capillary columns, DB-VRX
Supplier: Agilent
123-1564 122-1534 122-1564 123-1534 124-1534 124-1574 121-1524 121-1544
HEWL123-1564EA 1710 EUR
HEWL123-1564 HEWL122-1534 HEWL122-1564 HEWL123-1534 HEWL124-1534 HEWL124-1574 HEWL121-1524 HEWL121-1544
Capillary columns, DB-VRX
Chromatography Columns
DB-VRX has unique selectivity engineered for optimum resolution of volatiles analysis.
  • No subambient cooling required to resolve the six 'gases'
  • Fast run time
  • Low polarity
  • Excellent peak shape
  • Bonded and cross-linked
  • Solvent rinsable