Capillary columns, DB-5
Supplier: Agilent
121-5012 121-5013 121-5022 121-5023 121-5042 122-5011 122-5012 122-5013 122-501E 122-5022 122-5031 122-5032 122-5032E 122-5033 122-503E 122-5052 122-5061 122-5062 122-5063 122-506E 123-5011 123-5012 123-5012E 123-5013 123-5022 123-5026 123-502D 123-502F 123-5031 123-5032 123-5033 123-503B 123-503E 123-5052 123-5053 123-5056 123-5062 123-5063 124-5032 124-5037 125-5012 125-5017 125-501J 125-501K 125-5025 125-5032 125-5034 125-5035 125-5037 125-503B 125-503D 125-503J 125-503K 125-5062 125-5065 125-50HB 127-5012 127-5013 127-501E 127-501N 127-5022 127-5023 128-5012 128-5022 128-5052 128-50H7 12A-5015 123-5032E 125-5032E 123-5013E 123-5033E 125-5012E 125-5035E 125-5065E 127-5012E 127-5022E 121-5012E 121-5022E 122-5012E 122-5033E 122-5062E 122-5063E 123-500E 121-5012LTM 121-5013LTM 121-5022LTM 121-5023LTM 122-5002LTM 122-5012LTM 122-5032LTM 122-5033LTM 122-503ELTM 123-5003LTM 123-500ELTM 123-5011LTM 123-5012LTM 123-5013LTM 123-5032LTM 123-503BLTM 123-503ELTM 125-5032LTM 125-5035LTM 127-5012LTM 127-5013LTM 127-501ELTM 12A-5015LTM
HEWL121-5012EA 441 EUR
HEWL121-5012 HEWL121-5013 HEWL121-5022 HEWL121-5023 HEWL121-5042 HEWL122-5011 HEWL122-5012 HEWL122-5013 HEWL122-501E HEWL122-5022 HEWL122-5031 HEWL122-5032 HEWL122-5032E HEWL122-5033 HEWL122-503E HEWL122-5052 HEWL122-5061 HEWL122-5062 HEWL122-5063 HEWL122-506E HEWL123-5011 HEWL123-5012 HEWL123-5012E HEWL123-5013 HEWL123-5022 HEWL123-5026 HEWL123-502D HEWL123-502F HEWL123-5031 HEWL123-5032 HEWL123-5033 HEWL123-503B HEWL123-503E HEWL123-5052 HEWL123-5053 HEWL123-5056 HEWL123-5062 HEWL123-5063 HEWL124-5032 HEWL124-5037 HEWL125-5012 HEWL125-5017 HEWL125-501J HEWL125-501K HEWL125-5025 HEWL125-5032 HEWL125-5034 HEWL125-5035 HEWL125-5037 HEWL125-503B HEWL125-503D HEWL125-503J HEWL125-503K HEWL125-5062 HEWL125-5065 HEWL125-50HB HEWL127-5012 HEWL127-5013 HEWL127-501E HEWL127-501N HEWL127-5022 HEWL127-5023 HEWL128-5012 HEWL128-5022 HEWL128-5052 HEWL128-50H7 HEWL12A-5015 HEWL123-5032E HEWL125-5032E HEWL123-5013E HEWL123-5033E HEWL125-5012E HEWL125-5035E HEWL125-5065E HEWL127-5012E HEWL127-5022E HEWL121-5012E HEWL121-5022E HEWL122-5012E HEWL122-5033E HEWL122-5062E HEWL122-5063E HEWL123-500E HEWL121-5012LTM HEWL121-5013LTM HEWL121-5022LTM HEWL121-5023LTM HEWL122-5002LTM HEWL122-5012LTM HEWL122-5032LTM HEWL122-5033LTM HEWL122-503ELTM HEWL123-5003LTM HEWL123-500ELTM HEWL123-5011LTM HEWL123-5012LTM HEWL123-5013LTM HEWL123-5032LTM HEWL123-503BLTM HEWL123-503ELTM HEWL125-5032LTM HEWL125-5035LTM HEWL127-5012LTM HEWL127-5013LTM HEWL127-501ELTM HEWL12A-5015LTM
Capillary columns, DB-5
Chromatography Columns
These non-polar columns are used in wide range of applications.
  • Low bleed, bonded and cross-linked
  • Solvent rinsable
  • High temperature limit
(5%-Phenyl)-methylpolysiloxane.