Gas filters, charcoal
Supplier: Agilent
6810008600
HEWL6810008600EA 277 EUR
HEWL6810008600
Gas filters, charcoal
Filters Gas Filters
Gas filters are specifically designed to remove impurity gases, moisture and hydrocarbons from the ICP-OES gas supplies.
  • Deliver clean gases for accurate analyses
  • Both inlet and outlet connectors are equipped with stainless steel frits to prevent particulates from entering the gas stream
  • Easily replaced
This single combination filter removes water, trace impurity gases and organic compounds. No visual indicator. Installation date tag provided to provide a visual reminder when replacement required.