HPLC columns, Poroshell 120 EC-C8
Supplier: Agilent
699775-906 691775-906 691975-306 691975-906 693775-906 693975-306 693975-906 695775-906 695975-306 695975-906 697775-906 697975-306 697975-906 699975-306 699975-906 823750-913 821725-913 820750-913 699970-306 699970-906 695770-906 695970-306 695970-906 699770-906
HEWL699775-906EA 754 EUR
HEWL699775-906 HEWL691775-906 HEWL691975-306 HEWL691975-906 HEWL693775-906 HEWL693975-306 HEWL693975-906 HEWL695775-906 HEWL695975-306 HEWL695975-906 HEWL697775-906 HEWL697975-306 HEWL697975-906 HEWL699975-306 HEWL699975-906 HEWL823750-913 HEWL821725-913 HEWL820750-913 HEWL699970-306 HEWL699970-906 HEWL695770-906 HEWL695970-306 HEWL695970-906 HEWL699770-906
HPLC columns, Poroshell 120 EC-C8
Chromatography Columns
Poroshell 120 is made with a superficially porous particle that has a solid silica core and a porous outer layer. This particle provides high efficiency at lower pressures and is ideal for fast or high-resolution separations of many types of analytes.
  • Less retentive for non-polar samples
  • Exhaustive endcapping ideal for use with basic compounds, for those producing poor peak shapes on other columns
  • Ideal for a wide range of applications
The EC-C8 bonded phase is made by first chemically bonding a dense monolayer of dimethyl-n-octyl silane stationaryMore Product Information