Igniter supplies for PerkinElmer
Supplier: Agilent
8003-0381
HEWL8003-0381EA 10 EUR
HEWL8003-0381
Igniter supplies for PerkinElmer
Chromatography Supplies Chromatography Spare Parts
These igniter supplies are equivalent to the PerkinElmer ICP-OES versions for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Many parts in Agilent packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage.