HPLC columns, ZORBAX® Eclipse XDB
Supplier: Agilent
5064-8288 7995118-344 7995118-585 7995118-595 930990-902 931975-902 931975-906 931975-932 933975-902 933975-906 933975-932 935967-902 935967-912 946975-902 946975-906 960967-902 960967-912 963954-302 963954-306 963954-312 963967-902 963967-912 966967-902 966967-912 971700-902 973700-902 973700-906 973700-932 975700-902 975700-906 975700-932 990967-202 990967-302 990967-312 990967-902 990967-906 990967-912 993700-902 993700-912 993967-302 993967-306 993967-312 993967-902 993967-906 993967-912 7995230-344 961600-902 961967-902 963600-902 965600-902 922975-902 820555-901 923975-902 923700-932 5185-5921 921700-902 921700-932 921975-902 921975-932 922975-932 923700-902 923975-932 925700-902 925700-932 925975-932 961753-902 961753-905 961967-302 961967-905 963954-305 963967-905 966735-902 966954-302 966967-905 990967-305 990967-905 993700-905 993967-305 993967-905 960967-905 927700-902 928700-906 924700-902 924975-902 928700-902 927975-902 927975-906 974700-902 928975-902 927975-302 927975-306 928975-302 928975-306 924700-906 924975-302 924975-306 924975-906 926700-902 926700-906 926975-302 926975-306 926975-902 926975-906 927700-906 928975-906 932967-902 972700-902 981758-902 981757-302 981757-902 981758-302 981759-902 981759-302 823750-903 820750-903 820675-112 927975-902K 928975-902K 935967-902K 961967-902K 963967-902K 981757-302K 981758-302K 981759-302K 981759-902K 990967-902K 993967-902K
HEWL5064-8288EA 0 EUR
HEWL5064-8288 HEWL7995118-344 HEWL7995118-585 HEWL7995118-595 HEWL930990-902 HEWL931975-902 HEWL931975-906 HEWL931975-932 HEWL933975-902 HEWL933975-906 HEWL933975-932 HEWL935967-902 HEWL935967-912 HEWL946975-902 HEWL946975-906 554-1298 HEWL960967-912 HEWL963954-302 HEWL963954-306 HEWL963954-312 HEWL963967-902 HEWL963967-912 HEWL966967-902 HEWL966967-912 HEWL971700-902 HEWL973700-902 HEWL973700-906 HEWL973700-932 HEWL975700-902 HEWL975700-906 HEWL975700-932 HEWL990967-202 HEWL990967-302 HEWL990967-312 554-1299 554-1300 HEWL990967-912 HEWL993700-902 HEWL993700-912 HEWL993967-302 HEWL993967-306 HEWL993967-312 HEWL993967-902 554-1301 HEWL993967-912 HEWL7995230-344 HEWL961600-902 HEWL961967-902 HEWL963600-902 HEWL965600-902 HEWL922975-902 HEWL820555-901 HEWL923975-902 HEWL923700-932 HEWL5185-5921 HEWL921700-902 HEWL921700-932 HEWL921975-902 HEWL921975-932 HEWL922975-932 HEWL923700-902 HEWL923975-932 HEWL925700-902 HEWL925700-932 HEWL925975-932 HEWL961753-902 HEWL961753-905 HEWL961967-302 HEWL961967-905 HEWL963954-305 HEWL963967-905 HEWL966735-902 HEWL966954-302 HEWL966967-905 HEWL990967-305 HEWL990967-905 HEWL993700-905 HEWL993967-305 HEWL993967-905 HEWL960967-905 HEWL927700-902 HEWL928700-906 HEWL924700-902 HEWL924975-902 HEWL928700-902 HEWL927975-902 HEWL927975-906 HEWL974700-902 HEWL928975-902 HEWL927975-302 HEWL927975-306 HEWL928975-302 554-1429 HEWL924700-906 HEWL924975-302 HEWL924975-306 HEWL924975-906 HEWL926700-902 HEWL926700-906 HEWL926975-302 HEWL926975-306 HEWL926975-902 HEWL926975-906 HEWL927700-906 HEWL928975-906 HEWL932967-902 HEWL972700-902 HEWL981758-902 HEWL981757-302 HEWL981757-902 HEWL981758-302 HEWL981759-902 HEWL981759-302 HEWL823750-903 HEWL820750-903 HEWL820675-112 HEWL927975-902K HEWL928975-902K HEWL935967-902K HEWL961967-902K HEWL963967-902K HEWL981757-302K HEWL981758-302K HEWL981759-302K HEWL981759-902K HEWL990967-902K HEWL993967-902K
HPLC columns, ZORBAX® Eclipse XDB
Chromatography Columns
Good peak shape for basic, acidic and neutral compounds.
  • High performance over a wide pH range (2 to 9)
  • Long lifetime with extra dens bonding and double endcapping