Anti-CDC25C Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
13-986
PRSI13-986EA 419 EUR
PRSI13-986
Anti-CDC25C Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CDC25C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR