Anti-PTBP1 Rabbit Polyclonal Antibody

Supplier: ProSci Inc.
22-009
PRSI22-009EA 419 EUR
PRSI22-009
Anti-PTBP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PTBP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR